Product Details
- SNP ID
-
rs41313272
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:67093073 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAGTGGCGTTACATTATTTAGTT[C/T]ATATATATATTTAAAAGGCTTTGAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf141
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41299551,rs567792598] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf141
- Gene Name
- chromosome 1 open reading frame 141
View Full Product Details