Product Details

SNP ID
rs41459145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60852799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCAGCTTGCGCAGCATGTCGGG[G/T]CAGGTGAGCACGTAGAGCACCGGGT
Phenotype
MIM: 611293 MIM: 604837
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC86 PubMed Links

Gene Details

Gene
CCDC86
Gene Name
coiled-coil domain containing 86
There are no transcripts associated with this gene.

Gene
PTGDR2
Gene Name
prostaglandin D2 receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004778.2 1036 Silent Mutation NP_004769.2

View Full Product Details