Product Details

SNP ID
rs12797872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:833849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGTGGCAGACACATCCCCCCG[A/C]TGGTGGCAGGGTCTGCCTGTGCCAC
Phenotype
MIM: 602243 MIM: 614177 MIM: 609059 MIM: 601189 MIM: 602644
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD151 PubMed Links

Gene Details

Gene
CD151
Gene Name
CD151 molecule (Raph blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039490.1 Intron NP_001034579.1
NM_004357.4 Intron NP_004348.2
NM_139029.1 Intron NP_620598.1
NM_139030.3 Intron NP_620599.1
Gene
CRACR2B
Gene Name
calcium release activated channel regulator 2B
There are no transcripts associated with this gene.

Gene
PNPLA2
Gene Name
patatin like phospholipase domain containing 2
There are no transcripts associated with this gene.

Gene
POLR2L
Gene Name
RNA polymerase II subunit L
There are no transcripts associated with this gene.

Gene
TSPAN4
Gene Name
tetraspanin 4
There are no transcripts associated with this gene.

View Full Product Details