Product Details

SNP ID
rs60877936
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:1179670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCCTCCAGGGACCATCCCTGC[G/T]TCACGTCTGCACCCAGCACCGGCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TTLL10 PubMed Links

Gene Details

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130045.1 442 Silent Mutation GCG,GCT A44A NP_001123517.1
NM_153254.2 442 Intron NP_694986.2
XM_005244738.1 442 UTR 5 XP_005244795.1
XM_011541177.2 442 Silent Mutation GCG,GCT A44A XP_011539479.1
XM_017000906.1 442 Intron XP_016856395.1
XM_017000907.1 442 Silent Mutation GCG,GCT A44A XP_016856396.1
XM_017000908.1 442 Silent Mutation GCG,GCT A44A XP_016856397.1
XM_017000909.1 442 Silent Mutation GCG,GCT A44A XP_016856398.1
XM_017000910.1 442 Silent Mutation GCG,GCT A44A XP_016856399.1
XM_017000911.1 442 Silent Mutation GCG,GCT A44A XP_016856400.1
XM_017000912.1 442 Silent Mutation GCG,GCT A44A XP_016856401.1
Gene
TTLL10-AS1
Gene Name
TTLL10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details