Product Details
- SNP ID
-
rs55954760
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
19
- Location
-
Chr.1:150731739 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAGCAGTGGAGTATTGGTTATTT[C/T]ATTCTGGTTGGCTCAGTTATTTGCC
- Phenotype
-
MIM: 116845
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTSS
PubMed Links
Gene Details
- Gene
- CTSS
- Gene Name
- cathepsin S
View Full Product Details