Product Details

SNP ID
rs59879537
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:168246047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGTGACGCAATGAAGCATCCAG[C/T]AAAGCTTTTGTTGTTGATTGTTTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD36BP1 PubMed Links

Gene Details

Gene
ANKRD36BP1
Gene Name
ankyrin repeat domain 36B pseudogene 1
There are no transcripts associated with this gene.

Gene
SFT2D2
Gene Name
SFT2 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details