Product Details

SNP ID
rs61758743
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:156066092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGCTGGCCTGGAGCGGTACCG[A/C]GCCATCACCTCGGCGTGAGCCCGGG
Phenotype
MIM: 610389 MIM: 611007 MIM: 612942
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LAMTOR2 PubMed Links

Gene Details

Gene
LAMTOR2
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
There are no transcripts associated with this gene.

Gene
MEX3A
Gene Name
mex-3 RNA binding family member A
There are no transcripts associated with this gene.

Gene
RAB25
Gene Name
RAB25, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020387.3 477 Silent Mutation CGA,CGC R75R NP_065120.2

View Full Product Details