Product Details

SNP ID
rs58066571
Assay Type
Functionally Tested
NCBI dbSNP Submissions
21
Location
Chr.1:28900817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCCCAGGGACTTTGCACTTGC[C/T]AGTCCTTTGGCCCAGAACACTCTAC
Phenotype
MIM: 130500
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EPB41 PubMed Links

Gene Details

Gene
EPB41
Gene Name
erythrocyte membrane protein band 4.1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166005.1 Intron NP_001159477.1
NM_001166006.1 Intron NP_001159478.1
NM_001166007.1 Intron NP_001159479.1
NM_004437.3 Intron NP_004428.1
NM_203342.2 Intron NP_976217.1
NM_203343.2 Intron NP_976218.1
XM_005245753.1 Intron XP_005245810.1
XM_005245757.1 Intron XP_005245814.1
XM_005245760.1 Intron XP_005245817.1
XM_005245761.1 Intron XP_005245818.1
XM_005245763.1 Intron XP_005245820.1
XM_005245764.1 Intron XP_005245821.1
XM_005245765.1 Intron XP_005245822.1
XM_005245768.1 Intron XP_005245825.1
XM_005245769.1 Intron XP_005245826.1
XM_005245770.1 Intron XP_005245827.1
XM_005245772.4 Intron XP_005245829.1
XM_005245773.4 Intron XP_005245830.1
XM_005245774.1 Intron XP_005245831.1
XM_006710434.1 Intron XP_006710497.1
XM_006710439.1 Intron XP_006710502.1
XM_011540956.1 Intron XP_011539258.1
XM_011540957.1 Intron XP_011539259.1
XM_011540958.1 Intron XP_011539260.1
XM_011540959.1 Intron XP_011539261.1
XM_011540960.1 Intron XP_011539262.1
XM_011540961.1 Intron XP_011539263.1
XM_011540962.1 Intron XP_011539264.1
XM_011540963.2 Intron XP_011539265.1
XM_011540964.1 Intron XP_011539266.1
XM_011540965.2 Intron XP_011539267.1
XM_017000581.1 Intron XP_016856070.1
XM_017000582.1 Intron XP_016856071.1
XM_017000583.1 Intron XP_016856072.1
XM_017000584.1 Intron XP_016856073.1
XM_017000585.1 Intron XP_016856074.1
XM_017000586.1 Intron XP_016856075.1
XM_017000587.1 Intron XP_016856076.1
XM_017000588.1 Intron XP_016856077.1
XM_017000589.1 Intron XP_016856078.1
XM_017000590.1 Intron XP_016856079.1
XM_017000591.1 Intron XP_016856080.1
XM_017000592.1 Intron XP_016856081.1
XM_017000593.1 Intron XP_016856082.1
XM_017000594.1 Intron XP_016856083.1
XM_017000595.1 Intron XP_016856084.1
XM_017000596.1 Intron XP_016856085.1
XM_017000597.1 Intron XP_016856086.1
XM_017000598.1 Intron XP_016856087.1
XM_017000599.1 Intron XP_016856088.1
XM_017000600.1 Intron XP_016856089.1
XM_017000601.1 Intron XP_016856090.1
XM_017000602.1 Intron XP_016856091.1
XM_017000603.1 Intron XP_016856092.1
XM_017000604.1 Intron XP_016856093.1

View Full Product Details