Product Details

SNP ID
rs61019331
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:20055214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGATGAAACCCCCACACAGTCAC[A/G]GAATATTGTGTGCTGGATTGTTCCA
Phenotype
MIM: 601192
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G5 PubMed Links

Gene Details

Gene
PLA2G5
Gene Name
phospholipase A2 group V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000929.2 Intron NP_000920.1
XM_005245891.4 Intron XP_005245948.1
XM_005245892.4 Intron XP_005245949.1
XM_005245893.4 Intron XP_005245950.1
XM_011541586.2 Intron XP_011539888.1
XM_011541587.2 Intron XP_011539889.1
XM_011541588.2 Intron XP_011539890.1
XM_011541589.2 Intron XP_011539891.1
XM_011541590.2 Intron XP_011539892.1
XM_011541591.2 Intron XP_011539893.1
XM_011541592.2 Intron XP_011539894.1

View Full Product Details