Product Details

SNP ID
rs59717136
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:22030380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAATGATCAAATGAGATCACT[A/G]CCCAATGAAATGATGGGCCGAGGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00339 PubMed Links

Gene Details

Gene
LINC00339
Gene Name
long intergenic non-protein coding RNA 339
There are no transcripts associated with this gene.

Gene
LOC101928043
Gene Name
uncharacterized LOC101928043
There are no transcripts associated with this gene.

View Full Product Details