Product Details

SNP ID
hCV88409373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9474641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTTGCTCTTCGGGGACTCCAT[A/C]ACCCAGGTACGGCCGCCCCGACGCT
Phenotype
MIM: 606029
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPSF3 PubMed Links

Gene Details

Gene
CPSF3
Gene Name
cleavage and polyadenylation specific factor 3
There are no transcripts associated with this gene.

Gene
IAH1
Gene Name
isoamyl acetate-hydrolyzing esterase 1 homolog
There are no transcripts associated with this gene.

View Full Product Details