Product Details

SNP ID
rs1803686
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:109737464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGCAGGAAACGTCACCTCAAAA[C/A]GGCACATGAAAGCCTTCAGGTTTGG
Phenotype
MIM: 138390
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
GSTM3 PubMed Links

Gene Details

Gene
GSTM3
Gene Name
glutathione S-transferase mu 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000849.4 882 Missense Mutation NP_000840.2

View Full Product Details