Product Details

SNP ID
rs56224946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:159803909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAAAACAAAATTCTTAGTCAAA[A/G]GTTGACATGCTATGTGAAGCATATA
Phenotype
MIM: 612246 MIM: 604524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD302 PubMed Links

Gene Details

Gene
CD302
Gene Name
CD302 molecule
There are no transcripts associated with this gene.

Gene
LY75
Gene Name
lymphocyte antigen 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002349.3 6378 UTR 3 NP_002340.2
Gene
LY75-CD302
Gene Name
LY75-CD302 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198759.1 6378 Intron NP_001185688.1
NM_001198760.1 6378 Intron NP_001185689.1

View Full Product Details