Product Details

SNP ID
rs61752206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218164274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGGATCTTGAGGAATCCATGG[C/T]GAAAATTTTGGCCGATGAAGGCGTA
Phenotype
MIM: 146929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCR1 PubMed Links

Gene Details

Gene
CXCR1
Gene Name
C-X-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000634.2 1057 Missense Mutation CAC,CGC H313R NP_000625.1

View Full Product Details