Product Details

SNP ID
rs56008219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240886736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGCATTTCGTCACCAAGGAATG[G/T]CATACAGTGCAGGCAGGGCCAAGTC
Phenotype
MIM: 604285
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGXT PubMed Links

Gene Details

Gene
AGXT
Gene Name
alanine-glyoxylate aminotransferase
There are no transcripts associated with this gene.

Gene
C2orf54
Gene Name
chromosome 2 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085437.2 1837 UTR 3 NP_001078906.2
NM_001282921.1 1837 UTR 3 NP_001269850.1
NM_024861.3 1837 UTR 3 NP_079137.3
XM_011511877.1 1837 UTR 3 XP_011510179.1

View Full Product Details