Product Details

SNP ID
rs59492803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158702908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCTAGGTTTTTAGTTGATGTG[G/T]GTTGTACATGTGGTCTTTGGAAAGC
Phenotype
MIM: 605090
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RARRES1 PubMed Links

Gene Details

Gene
RARRES1
Gene Name
retinoic acid receptor responder 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002888.3 Intron NP_002879.2
NM_206963.1 Intron NP_996846.1
XM_005247686.4 Intron XP_005247743.3

View Full Product Details