Product Details

SNP ID
rs59790563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101563553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATAGATTGAAAAAAAAGAATAG[A/G]ATAGAATGCGGAGTTACATCCACCT
Phenotype
MIM: 615423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRMT10C PubMed Links

Gene Details

Gene
TRMT10C
Gene Name
tRNA methyltransferase 10C, mitochondrial RNase P subunit
There are no transcripts associated with this gene.

View Full Product Details