Product Details

SNP ID
rs55996736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:660925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATGATGCCAGCCCAACTCCAC[C/T]ATCTGCGCCCAAACTTGGACAGGAT
Phenotype
MIM: 616475 MIM: 608773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP72 PubMed Links

Gene Details

Gene
CEP72
Gene Name
centrosomal protein 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018140.3 7155 Intron NP_060610.2
XM_005248322.3 7155 Intron XP_005248379.1
XM_011514063.1 7155 Intron XP_011512365.1
XM_011514064.2 7155 Intron XP_011512366.1
XM_017009626.1 7155 Intron XP_016865115.1
XM_017009627.1 7155 Intron XP_016865116.1
Gene
LOC101929895
Gene Name
uncharacterized LOC101929895
There are no transcripts associated with this gene.

Gene
TPPP
Gene Name
tubulin polymerization promoting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007030.2 7155 UTR 3 NP_008961.1
XM_005248237.3 7155 UTR 3 XP_005248294.2
XM_017008993.1 7155 UTR 3 XP_016864482.1
XM_017008994.1 7155 UTR 3 XP_016864483.1

View Full Product Details