Product Details

SNP ID
rs60168628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:162728210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGATTCTTTTACTACACTTTTT[A/C]TGAGAACAAGACATTTTCTAGGAAG
Phenotype
MIM: 608427 MIM: 602544
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PACRG PubMed Links

Gene Details

Gene
PACRG
Gene Name
PARK2 coregulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080378.1 124 UTR 5 NP_001073847.1
NM_001080379.1 124 UTR 5 NP_001073848.1
NM_152410.2 124 UTR 5 NP_689623.2
XM_005266825.4 124 UTR 5 XP_005266882.1
XM_005266826.4 124 UTR 5 XP_005266883.1
XM_006715344.3 124 UTR 5 XP_006715407.1
XM_006715345.3 124 UTR 5 XP_006715408.1
XM_011535461.2 124 UTR 5 XP_011533763.1
XM_011535462.2 124 UTR 5 XP_011533764.1
XM_011535466.1 124 UTR 5 XP_011533768.1
XM_011535468.2 124 UTR 5 XP_011533770.1
XM_011535469.2 124 UTR 5 XP_011533771.1
XM_011535471.1 124 UTR 5 XP_011533773.1
XM_017010275.1 124 UTR 5 XP_016865764.1
XM_017010276.1 124 UTR 5 XP_016865765.1
XM_017010277.1 124 UTR 5 XP_016865766.1
XM_017010278.1 124 UTR 5 XP_016865767.1
XM_017010279.1 124 UTR 5 XP_016865768.1
XM_017010280.1 124 UTR 5 XP_016865769.1
XM_017010281.1 124 Intron XP_016865770.1
XM_017010282.1 124 Intron XP_016865771.1
Gene
PARK2
Gene Name
parkin RBR E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004562.2 124 Intron NP_004553.2
NM_013987.2 124 Intron NP_054642.2
NM_013988.2 124 Intron NP_054643.2
XM_011535863.1 124 Intron XP_011534165.1
XM_017010908.1 124 Intron XP_016866397.1
XM_017010909.1 124 Intron XP_016866398.1

View Full Product Details