Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002114.3 | 214 | Intron | NP_002105.3 | ||
XM_011514546.2 | 214 | UTR 5 | XP_011512848.1 | ||
XM_011514547.2 | 214 | Intron | XP_011512849.1 | ||
XM_011514548.2 | 214 | UTR 5 | XP_011512850.1 | ||
XM_011514549.2 | 214 | Intron | XP_011512851.1 | ||
XM_011514550.2 | 214 | Intron | XP_011512852.1 | ||
XM_011514551.2 | 214 | UTR 5 | XP_011512853.1 | ||
XM_011514552.2 | 214 | Intron | XP_011512854.1 | ||
XM_011514553.1 | 214 | Intron | XP_011512855.1 | ||
XM_011514555.2 | 214 | Intron | XP_011512857.1 | ||
XM_017010800.1 | 214 | Intron | XP_016866289.1 | ||
XM_017010801.1 | 214 | Intron | XP_016866290.1 | ||
XM_017010802.1 | 214 | Intron | XP_016866291.1 | ||
XM_017010803.1 | 214 | UTR 5 | XP_016866292.1 | ||
XM_017010804.1 | 214 | UTR 5 | XP_016866293.1 |