Product Details
- SNP ID
-
rs61756281
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:14149164 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTTCGGTTTCTTGTCCTTTTGAC[A/G]AGGGAGCAGAATAAACCGGTTTTTG
- Phenotype
-
MIM: 604070
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DGKB
PubMed Links
Gene Details
- Gene
- DGKB
- Gene Name
- diacylglycerol kinase beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004080.2 |
3094 |
Silent Mutation |
CTC,CTT |
L794L |
NP_004071.1 |
NM_145695.2 |
3094 |
Intron |
|
|
NP_663733.1 |
XM_005249628.2 |
3094 |
Silent Mutation |
CTC,CTT |
L794L |
XP_005249685.1 |
XM_005249629.2 |
3094 |
Silent Mutation |
CTC,CTT |
L793L |
XP_005249686.1 |
XM_005249630.2 |
3094 |
Silent Mutation |
CTC,CTT |
L787L |
XP_005249687.1 |
XM_005249631.3 |
3094 |
Silent Mutation |
CTC,CTT |
L775L |
XP_005249688.1 |
XM_011515153.2 |
3094 |
Silent Mutation |
CTC,CTT |
L794L |
XP_011513455.1 |
XM_011515154.2 |
3094 |
Silent Mutation |
CTC,CTT |
L794L |
XP_011513456.1 |
XM_011515156.2 |
3094 |
Silent Mutation |
CTC,CTT |
L786L |
XP_011513458.1 |
XM_011515157.2 |
3094 |
Silent Mutation |
CTC,CTT |
L782L |
XP_011513459.1 |
XM_011515158.2 |
3094 |
Silent Mutation |
CTC,CTT |
L775L |
XP_011513460.1 |
XM_017011783.1 |
3094 |
Silent Mutation |
CTC,CTT |
L793L |
XP_016867272.1 |
XM_017011784.1 |
3094 |
Silent Mutation |
CTC,CTT |
L793L |
XP_016867273.1 |
XM_017011785.1 |
3094 |
Silent Mutation |
CTC,CTT |
L787L |
XP_016867274.1 |
XM_017011786.1 |
3094 |
Silent Mutation |
CTC,CTT |
L786L |
XP_016867275.1 |
XM_017011787.1 |
3094 |
Silent Mutation |
CTC,CTT |
L786L |
XP_016867276.1 |
XM_017011788.1 |
3094 |
Silent Mutation |
CTC,CTT |
L782L |
XP_016867277.1 |
XM_017011789.1 |
3094 |
Silent Mutation |
CTC,CTT |
L782L |
XP_016867278.1 |
XM_017011790.1 |
3094 |
Intron |
|
|
XP_016867279.1 |
XM_017011791.1 |
3094 |
Intron |
|
|
XP_016867280.1 |
XM_017011792.1 |
3094 |
Intron |
|
|
XP_016867281.1 |
View Full Product Details