Product Details

SNP ID
rs62456178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5990656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCTAGAACACAATACAGAAAAA[A/G]CCTTAAAATGATCATATTCACTGAC
Phenotype
MIM: 600259
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PMS2 PubMed Links

Gene Details

Gene
PMS2
Gene Name
PMS1 homolog 2, mismatch repair system component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000535.6 Intron NP_000526.2
NM_001322003.1 Intron NP_001308932.1
NM_001322004.1 Intron NP_001308933.1
NM_001322005.1 Intron NP_001308934.1
NM_001322006.1 Intron NP_001308935.1
NM_001322007.1 Intron NP_001308936.1
NM_001322008.1 Intron NP_001308937.1
NM_001322009.1 Intron NP_001308938.1
NM_001322010.1 Intron NP_001308939.1
NM_001322011.1 Intron NP_001308940.1
NM_001322012.1 Intron NP_001308941.1
NM_001322013.1 Intron NP_001308942.1
NM_001322014.1 Intron NP_001308943.1
NM_001322015.1 Intron NP_001308944.1
XM_006715744.3 Intron XP_006715807.1
XM_017012342.1 Intron XP_016867831.1

View Full Product Details