Product Details

SNP ID
rs58107700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6621598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCAGCAAGTGCAAGAGCAACAG[C/G]TGTTACAGCAACAGCAGGAACAGTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF853 PubMed Links

Gene Details

Gene
ZNF853
Gene Name
zinc finger protein 853
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017560.1 1048 Missense Mutation CTG,GTG L203V NP_060030.1
XM_011515438.2 1048 Missense Mutation CTG,GTG L229V XP_011513740.1
XM_011515439.2 1048 Missense Mutation CTG,GTG L214V XP_011513741.1
XM_017012371.1 1048 Missense Mutation CTG,GTG L188V XP_016867860.1

View Full Product Details