Product Details

SNP ID
rs60344499
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143935266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAACCAGACGTGGGTGAGAGAA[A/T]TTATTCTCCTTGGCTTATCCAGTGA
Phenotype
MIM: 608497
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR2F1 PubMed Links
Additional Information
For this assay, SNP(s) [rs2240358] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR2F1
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012369.2 34 Intron NP_036501.2
XM_011516019.2 34 Intron XP_011514321.1
XM_011516020.2 34 Intron XP_011514322.1
Gene
OR2F2
Gene Name
olfactory receptor family 2 subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004685.1 34 Missense Mutation ATT,TTT I12F NP_001004685.1

View Full Product Details