Product Details

SNP ID
rs1062771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140456654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGACTGGGGAGGAGTCTCACTT[G/T]ATGATCTTGCTCTCAAATTGCTTAG
Phenotype
MIM: 607754
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MKRN1 PubMed Links

Gene Details

Gene
MKRN1
Gene Name
makorin ring finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145125.1 998 Silent Mutation ATA,ATC I328I NP_001138597.1
NM_001291663.1 998 Silent Mutation ATA,ATC I264I NP_001278592.1
NM_013446.3 998 Silent Mutation ATA,ATC I328I NP_038474.2
XM_011515996.1 998 Silent Mutation ATA,ATC I285I XP_011514298.1
XM_011515997.2 998 Silent Mutation ATA,ATC I264I XP_011514299.1
XM_011515998.1 998 Intron XP_011514300.1

View Full Product Details