Product Details

SNP ID
rs56928841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:86878631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTCCAGTCTATACAGCTTTGC[C/T]AGGGTTCCTCTGAAAGGGTTCATTT
Phenotype
MIM: 614048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1324L PubMed Links

Gene Details

Gene
KIAA1324L
Gene Name
KIAA1324 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142749.2 5115 UTR 3 NP_001136221.1
NM_001291990.1 5115 UTR 3 NP_001278919.1
NM_001291991.1 5115 UTR 3 NP_001278920.1
NM_152748.3 5115 UTR 3 NP_689961.3
XM_006715894.3 5115 UTR 3 XP_006715957.1
XM_011515918.2 5115 UTR 3 XP_011514220.1
XM_011515921.1 5115 UTR 3 XP_011514223.1
XM_017011843.1 5115 UTR 3 XP_016867332.1
XM_017011844.1 5115 UTR 3 XP_016867333.1

View Full Product Details