Product Details

SNP ID
rs59268053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100680542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAAAAGCGCATCATATCAGACC[C/T]GGAAGGCCACCTGCTCCTCTCCCAT
Phenotype
MIM: 612064 MIM: 139390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIGYF1 PubMed Links

Gene Details

Gene
GIGYF1
Gene Name
GRB10 interacting GYF protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022574.4 5075 UTR 3 NP_072096.2
XM_005250532.1 5075 UTR 3 XP_005250589.1
XM_011516472.1 5075 UTR 3 XP_011514774.1
XM_011516477.1 5075 UTR 3 XP_011514779.1
XM_017012526.1 5075 UTR 3 XP_016868015.1
XM_017012527.1 5075 UTR 3 XP_016868016.1
XM_017012528.1 5075 UTR 3 XP_016868017.1
XM_017012529.1 5075 UTR 3 XP_016868018.1
XM_017012530.1 5075 UTR 3 XP_016868019.1
XM_017012531.1 5075 UTR 3 XP_016868020.1
Gene
GNB2
Gene Name
G protein subunit beta 2
There are no transcripts associated with this gene.

View Full Product Details