Product Details

SNP ID
rs58307701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38516440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATAGAAATGGAAGAATGAGAGA[A/T]CACTGTGGGCTGCTGTGTTTTTGAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C8orf86 PubMed Links

Gene Details

Gene
C8orf86
Gene Name
chromosome 8 open reading frame 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303531.1 413 Intron NP_001290460.1
NM_207412.2 413 Missense Mutation ATC,TTC I130F NP_997295.1

View Full Product Details