Product Details

SNP ID
rs61752811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142873050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTCTGATGTTTGTCAGCAACCC[A/G]GTCTATGGCGTTTTCTTAGAGCATC
Phenotype
MIM: 610613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP11B1 PubMed Links

Gene Details

Gene
CYP11B1
Gene Name
cytochrome P450 family 11 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000497.3 2337 UTR 3 NP_000488.3
NM_001026213.1 2337 UTR 3 NP_001021384.1
XM_011516874.2 2337 UTR 3 XP_011515176.1
XM_011516875.1 2337 UTR 3 XP_011515177.1
XM_017013146.1 2337 UTR 3 XP_016868635.1

View Full Product Details