Product Details

SNP ID
rs61086334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:60189778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAACTATGAATCCTAGCAGCAG[A/T]AATGTGGATATATAATATAGGGTTT
Phenotype
MIM: 114815
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CA8 PubMed Links

Gene Details

Gene
CA8
Gene Name
carbonic anhydrase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321837.1 1457 Intron NP_001308766.1
NM_001321838.1 1457 Intron NP_001308767.1
NM_001321839.1 1457 UTR 3 NP_001308768.1
NM_004056.5 1457 UTR 3 NP_004047.3
XM_011517587.2 1457 Intron XP_011515889.1
XM_011517588.2 1457 Intron XP_011515890.1
XM_017013818.1 1457 UTR 3 XP_016869307.1

View Full Product Details