Product Details
- SNP ID
-
rs59308818
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:6013845 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGTCTTCAACAACAGCTGCATC[A/T]CATTTGATGCATCTTGCATAAATTT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA2026
PubMed Links
Gene Details
- Gene
- KIAA2026
- Gene Name
- KIAA2026
There are no transcripts associated with this gene.
- Gene
- MIR4665
- Gene Name
- microRNA 4665
There are no transcripts associated with this gene.
- Gene
- RANBP6
- Gene Name
- RAN binding protein 6
View Full Product Details