Product Details

SNP ID
rs55665019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128705503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATGGCGAGCTGCGGGAGCTGCA[C/T]GCCCGAATCCTGCTGCCCGGCCCTG
Phenotype
MIM: 610714 MIM: 600960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKN3 PubMed Links

Gene Details

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 708 Silent Mutation CAC,CAT H75H NP_001304855.1
NM_013355.4 708 Silent Mutation CAC,CAT H75H NP_037487.2
XM_005251946.3 708 Silent Mutation CAC,CAT H72H XP_005252003.1
XM_006717080.2 708 Silent Mutation CAC,CAT H82H XP_006717143.1
XM_017014649.1 708 Silent Mutation CAC,CAT H82H XP_016870138.1
XM_017014650.1 708 UTR 5 XP_016870139.1
Gene
SET
Gene Name
SET nuclear proto-oncogene
There are no transcripts associated with this gene.

View Full Product Details