Product Details

SNP ID
rs60499252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:60869805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATTAAGATGGATGCATTTGTA[A/G]CTATCCCTACTCTACTGCCTTCTCT
Phenotype
MIM: 607351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHOBTB1 PubMed Links

Gene Details

Gene
RHOBTB1
Gene Name
Rho related BTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242359.1 3982 UTR 3 NP_001229288.1
NM_014836.4 3982 UTR 3 NP_055651.1
XM_006718084.1 3982 UTR 3 XP_006718147.1
XM_006718085.1 3982 UTR 3 XP_006718148.1
XM_011540424.1 3982 UTR 3 XP_011538726.1
XM_011540425.1 3982 UTR 3 XP_011538727.1
XM_011540426.1 3982 Intron XP_011538728.1
XM_011540428.1 3982 Intron XP_011538730.1
XM_017017000.1 3982 UTR 3 XP_016872489.1
XM_017017001.1 3982 UTR 3 XP_016872490.1
XM_017017002.1 3982 UTR 3 XP_016872491.1
XM_017017003.1 3982 UTR 3 XP_016872492.1
XM_017017004.1 3982 Intron XP_016872493.1
XM_017017005.1 3982 UTR 3 XP_016872494.1
XM_017017006.1 3982 UTR 3 XP_016872495.1
XM_017017007.1 3982 UTR 3 XP_016872496.1
XM_017017008.1 3982 UTR 3 XP_016872497.1

View Full Product Details