Product Details
- SNP ID
-
rs60771337
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:75094679 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGATCCTGGCTGCCTGCCAGATCA[G/T]AACCGCCCCGTCTCCCGCCCCAGTC
- Phenotype
-
MIM: 613191
MIM: 611575
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DUSP13
PubMed Links
Gene Details
- Gene
- DUSP13
- Gene Name
- dual specificity phosphatase 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007271.1 |
1098 |
UTR 3 |
|
|
NP_001007272.1 |
NM_001007272.1 |
1098 |
Missense Mutation |
TTA,TTC |
L248F |
NP_001007273.1 |
NM_001007273.1 |
1098 |
Missense Mutation |
TTA,TTC |
L291F |
NP_001007274.1 |
NM_001320842.1 |
1098 |
Missense Mutation |
TTA,TTC |
L291F |
NP_001307771.1 |
NM_001320843.1 |
1098 |
Missense Mutation |
TTA,TTC |
L198F |
NP_001307772.1 |
NM_016364.3 |
1098 |
Missense Mutation |
TTA,TTC |
L198F |
NP_057448.3 |
XM_005269883.3 |
1098 |
Missense Mutation |
TTA,TTC |
L417F |
XP_005269940.1 |
XM_005269884.4 |
1098 |
Missense Mutation |
TTA,TTC |
L334F |
XP_005269941.3 |
XM_005269887.1 |
1098 |
Missense Mutation |
TTA,TTC |
L291F |
XP_005269944.1 |
XM_005269890.1 |
1098 |
Missense Mutation |
TTA,TTC |
L198F |
XP_005269947.1 |
XM_011539853.1 |
1098 |
Missense Mutation |
TTA,TTC |
L327F |
XP_011538155.1 |
XM_011539854.2 |
1098 |
Missense Mutation |
TTA,TTC |
L198F |
XP_011538156.1 |
XM_011539855.1 |
1098 |
Missense Mutation |
TTA,TTC |
L198F |
XP_011538157.1 |
XM_011539856.2 |
1098 |
Missense Mutation |
TTA,TTC |
L198F |
XP_011538158.1 |
XM_017016313.1 |
1098 |
Missense Mutation |
TTA,TTC |
L246F |
XP_016871802.1 |
XM_017016314.1 |
1098 |
Intron |
|
|
XP_016871803.1 |
- Gene
- SAMD8
- Gene Name
- sterile alpha motif domain containing 8
There are no transcripts associated with this gene.
View Full Product Details