Product Details

SNP ID
rs61757082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:58196509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTGGACAAAAACTTTTCTGCC[A/G]AAGTTCTGTCATTCAATTTTGTAGT
Phenotype
MIM: 609851
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPMK PubMed Links

Gene Details

Gene
IPMK
Gene Name
inositol polyphosphate multikinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152230.4 808 Intron NP_689416.1
XM_011539565.2 808 Intron XP_011537867.1
XM_017016013.1 808 Missense Mutation TCG,TTG S185L XP_016871502.1

View Full Product Details