Product Details

SNP ID
rs61749169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:28681328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATATGTGTAAATCTGAGCTCAG[C/T]GCCTGCTTCTCTAGACTTCTTGATC
Phenotype
MIM: 604444
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAMBI PubMed Links

Gene Details

Gene
BAMBI
Gene Name
BMP and activin membrane bound inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012342.2 550 Silent Mutation AGC,AGT S49S NP_036474.1

View Full Product Details