Product Details

SNP ID
rs61876191
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCACCCATTAAGATCAACATA[A/G]TCTATGGGCTCATGGTGATTTCTTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 595 Missense Mutation ATC,GTC I199V NP_001005165.1

View Full Product Details