Product Details

SNP ID
rs60116285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6891843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTGACCACAGTTACTATGATAC[C/G]GCCATAGGATACCAGAATCAGAAAA
Phenotype
MIM: 608494
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2D2 PubMed Links

Gene Details

Gene
OR2D2
Gene Name
olfactory receptor family 2 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003700.1 658 Missense Mutation CGT,GGT R220G NP_003691.1

View Full Product Details