Product Details

SNP ID
rs58115104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10455372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGTGGGTCACTTGAGGCAGGC[G/T]CCACCTTCCCCAGGAGGAGTGGCAG
Phenotype
MIM: 102772
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AMPD3 PubMed Links

Gene Details

Gene
AMPD3
Gene Name
adenosine monophosphate deaminase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000480.2 254 Intron NP_000471.1
NM_001025389.1 254 UTR 5 NP_001020560.1
NM_001025390.1 254 Intron NP_001020561.1
NM_001172430.1 254 Intron NP_001165901.1
NM_001172431.1 254 Intron NP_001165902.1

View Full Product Details