Product Details

SNP ID
rs59990432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12734468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGGCTATGGGAGACCACTGTAT[C/T]GATTCTTTCCATTTTGCAGATGAGA
Phenotype
MIM: 612456
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOLD1 PubMed Links

Gene Details

Gene
APOLD1
Gene Name
apolipoprotein L domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130415.1 Intron NP_001123887.1
NM_030817.2 Intron NP_110444.3

View Full Product Details