Product Details

SNP ID
rs58724135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116745062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTATTAATATTATCATTATTAT[C/T]ATCCAGAGCCCAGTGGAGGAGGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf49 PubMed Links

Gene Details

Gene
C12orf49
Gene Name
chromosome 12 open reading frame 49
There are no transcripts associated with this gene.

Gene
RNFT2
Gene Name
ring finger protein, transmembrane 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109903.1 Intron NP_001103373.1
NM_032814.3 Intron NP_116203.2

View Full Product Details