Product Details
- SNP ID
-
hCV89212109
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:63155628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATGCCAAATGATATACCTTCTGT[C/G]AGTCTGGCCTTTCCTCCTGTAGGCT
- Phenotype
-
MIM: 612055
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPS27L
PubMed Links
Gene Details
- Gene
- RPS27L
- Gene Name
- ribosomal protein S27 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015920.3 |
356 |
Silent Mutation |
CTC,CTG |
L73L |
NP_057004.1 |
View Full Product Details