Product Details

SNP ID
rs55720833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75607299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTACCTCATTGTAAATGCAAT[A/C]TAGAATGGTGTAGTCTGAGGACACA
Phenotype
MIM: 607902
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNUPN PubMed Links

Gene Details

Gene
SNUPN
Gene Name
snurportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042581.1 662 Missense Mutation GAT,TAT D173Y NP_001036046.1
NM_001042588.1 662 Missense Mutation GAT,TAT D173Y NP_001036053.1
NM_005701.3 662 Missense Mutation GAT,TAT D173Y NP_005692.1

View Full Product Details