Product Details

SNP ID
rs56160046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:38488655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATAAAAACTAACTGGGGATCTT[C/T]ACTTCGAATGGAGAGGAGGAAGAAT
Phenotype
MIM: 616142 MIM: 603962
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM98B PubMed Links

Gene Details

Gene
FAM98B
Gene Name
family with sequence similarity 98 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173611.3 4307 Intron NP_775882.2
Gene
RASGRP1
Gene Name
RAS guanyl releasing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128602.1 4307 UTR 3 NP_001122074.1
NM_001306086.1 4307 UTR 3 NP_001293015.1
NM_005739.3 4307 UTR 3 NP_005730.2
XM_005254114.2 4307 UTR 3 XP_005254171.1
XM_011521151.2 4307 UTR 3 XP_011519453.1
XM_017021860.1 4307 UTR 3 XP_016877349.1

View Full Product Details