Product Details

SNP ID
rs62625283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23603602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACACTGACCGAGAAGTAAGTCCC[A/C]AATGGCAATTGTTCCAGAAGTCAAG
Phenotype
MIM: 603836 MIM: 610355
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NDUFAB1 PubMed Links

Gene Details

Gene
NDUFAB1
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
There are no transcripts associated with this gene.

Gene
PALB2
Gene Name
partner and localizer of BRCA2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024675.3 4151 Missense Mutation GGG,TGG G1140W NP_078951.2
XM_011545946.2 4151 Missense Mutation GGG,TGG G1142W XP_011544248.1
XM_011545947.2 4151 UTR 3 XP_011544249.1
XM_011545948.2 4151 Missense Mutation GGG,TGG G845W XP_011544250.1
XM_017023671.1 4151 Missense Mutation GGG,TGG G1063W XP_016879160.1
XM_017023672.1 4151 Missense Mutation GGG,TGG G1061W XP_016879161.1
XM_017023673.1 4151 UTR 3 XP_016879162.1

View Full Product Details