Product Details

SNP ID
rs62029147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27791320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTCTGGTGGTCTTGCAGCAGG[A/G]GCTGGTGCCAGCGATGGCCTGAGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSG1L PubMed Links

Gene Details

Gene
GSG1L
Gene Name
GSG1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109763.1 978 UTR 3 NP_001103233.1
NM_001323900.1 978 UTR 3 NP_001310829.1
NM_001323901.1 978 UTR 3 NP_001310830.1
NM_144675.2 978 UTR 3 NP_653276.1
XM_011545750.2 978 Intron XP_011544052.1
XM_011545751.2 978 Intron XP_011544053.1

View Full Product Details