Product Details

SNP ID
rs61332682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75889623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATGAAGGTCCTGCCAGTGTCA[C/T]GTGCTGCTTAGTGACAGGGTTGAGG
Phenotype
MIM: 611844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL38 PubMed Links

Gene Details

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.

Gene
TRIM65
Gene Name
tripartite motif containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256124.1 2747 UTR 3 NP_001243053.1
NM_173547.3 2747 UTR 3 NP_775818.2
XM_006721760.3 2747 UTR 3 XP_006721823.1
XM_006721761.3 2747 UTR 3 XP_006721824.1
XM_011524499.2 2747 Intron XP_011522801.1
XM_011524500.2 2747 Intron XP_011522802.1
XM_011524501.2 2747 Intron XP_011522803.1
XM_011524502.2 2747 Intron XP_011522804.1
XM_011524503.2 2747 Intron XP_011522805.1
XM_017024343.1 2747 Intron XP_016879832.1
XM_017024344.1 2747 UTR 3 XP_016879833.1

View Full Product Details