Product Details
- SNP ID
-
rs59986127
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82528797 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAAGGTGCTCTGCTGAGAATTC[C/T]GTAGATTTCTAGACAGTATTTGGTT
- Phenotype
-
MIM: 147685
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXK2
PubMed Links
Gene Details
- Gene
- FOXK2
- Gene Name
- forkhead box K2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004514.3 |
|
Intron |
|
|
NP_004505.2 |
View Full Product Details