Product Details

SNP ID
rs61479360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4774267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTATATCACAGGGACACAGACAG[G/T]TTCCTGCTCTCCCTGAAGCTTGTGT
Phenotype
MIM: 604657
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TM4SF5 PubMed Links

Gene Details

Gene
TM4SF5
Gene Name
transmembrane 4 L six family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003963.2 Intron NP_003954.2

View Full Product Details