Product Details

SNP ID
rs61754327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42780995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGCCCCGCGAGGAGCCCACC[G/T]CCTAGCTCCAAAGAACCCCCCGAGG
Phenotype
MIM: 610907 MIM: 601844
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VPS25 PubMed Links

Gene Details

Gene
VPS25
Gene Name
vacuolar protein sorting 25 homolog
There are no transcripts associated with this gene.

Gene
WNK4
Gene Name
WNK lysine deficient protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321299.1 365 UTR 5 NP_001308228.1
NM_032387.4 365 Silent Mutation CCG,CCT P99P NP_115763.2
XM_011525135.2 365 Silent Mutation CCG,CCT P99P XP_011523437.1
XM_017024962.1 365 Silent Mutation CCG,CCT P99P XP_016880451.1
XM_017024963.1 365 Silent Mutation CCG,CCT P99P XP_016880452.1
XM_017024964.1 365 Silent Mutation CCG,CCT P99P XP_016880453.1
XM_017024965.1 365 Silent Mutation CCG,CCT P99P XP_016880454.1
XM_017024966.1 365 Intron XP_016880455.1

View Full Product Details